missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 113.00 - € 363.00
Specifications
| Antigen | CRTAP |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18631294
|
Novus Biologicals
NBP3-21432-25ul |
25 μg |
€ 113.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651235
|
Novus Biologicals
NBP3-21432-100ul |
100 μg |
€ 363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CRTAP Polyclonal antibody specifically detects CRTAP in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| CRTAP | |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH 7.0 | |
| 10491 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| cartilage associated protein, cartilage-associated protein, CASPLEPREL3, leprecan-like 3 | |
| Produced in rabbits immunized with E. coli-derived Human CRTAP fragment | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title