missing translation for 'onlineSavingsMsg'
Learn More

Tempshield™ Cryo-Gloves™ Blue Waterproof Mid-Arm Length Cryogenic Gloves

Product Code. 10248904
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
1 pair
Each
This item is not returnable. View return policy
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.

Product Code. 10248904

Brand: Tempshield™ MAMWP

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.

A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).

A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).

  • Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
  • Thin, seamless 100% waterproof liner provides protection from spills and splashes
  • State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
  • High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
  • Ideal for applications where function is important and safety is critical
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids

Specifications

Certifications/Compliance (Europe) EN 420, EN 511
Description Mid-Arm Length
Color Blue
Hand Specific
Product Type Waterproof Cryogenic Gloves
Size Medium
EN388 Standard Yes
For Use With (Application) Dispensing/Transferring Cryogenic Liquids, Removing Samples from Cryogenic Liquids, and Open Cryogenic Containers with the Chance of Exposure to Liquids/Splashes
Series Cryo-Gloves™
Temperature (English) Min -320°F
Temperature (Metric) Min. -196°C
Length (English) 13.3 to 15.3 in.
Length (Metric) 33.5 to 39.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.