missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSAD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35392-20ul
This item is not returnable.
View return policy
Description
CSAD Polyclonal antibody specifically detects CSAD in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| CSAD | |
| Polyclonal | |
| Western Blot 1:2000 - 1:6000, ELISA | |
| CSDMGC119355, cysteine sulfinic acid decarboxylase, Cysteine-sulfinate decarboxylase, EC 4.1.1, EC 4.1.1.29, FLJ44987, FLJ45500, MGC119354, MGC119357, PCAP, P-selectin cytoplasmic tail-associated protein, Sulfinoalanine decarboxylase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 371-520 of human CSAD (NP_057073.4).,, Sequence:, DVALDTGDKVVQCGRRVDCLKLWLMWKAQGDQGLERRIDQAFVLARYLVEEMKKREGFELVMEPEFVNVCFWFVPPSLRGKQESPDYHERLSKVAPVLKERMVKEGSMMIGYQPHGTRGNFFRVVVANSALTCADMDFLLNELERLGQDL | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51380 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction