missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSAD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | CSAD |
|---|---|
| Dilution | Western Blot 1:2000 - 1:6000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230418
|
Novus Biologicals
NBP3-35392-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228715
|
Novus Biologicals
NBP3-35392-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CSAD Polyclonal antibody specifically detects CSAD in Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| CSAD | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| CSDMGC119355, cysteine sulfinic acid decarboxylase, Cysteine-sulfinate decarboxylase, EC 4.1.1, EC 4.1.1.29, FLJ44987, FLJ45500, MGC119354, MGC119357, PCAP, P-selectin cytoplasmic tail-associated protein, Sulfinoalanine decarboxylase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 371-520 of human CSAD (NP_057073.4).,, Sequence:, DVALDTGDKVVQCGRRVDCLKLWLMWKAQGDQGLERRIDQAFVLARYLVEEMKKREGFELVMEPEFVNVCFWFVPPSLRGKQESPDYHERLSKVAPVLKERMVKEGSMMIGYQPHGTRGNFFRVVVANSALTCADMDFLLNELERLGQDL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:2000 - 1:6000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 51380 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title