missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSE1L Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22219-1-AP-150UL
This item is not returnable.
View return policy
Description
Proteins that carry a nuclear localization signal (NLS) are transported into the nucleus by the importin-alpha/beta heterodimer. Importin-alpha binds the NLS, while importin-beta mediates translocation through the nuclear pore complex. After translocation, RanGTP binds importin-beta and displaces importin-alpha. Importin-alpha must then be returned to the cytoplasm, leaving the NLS protein behind. The protein encoded by this gene binds strongly to NLS-free importin-alpha, and this binding is released in the cytoplasm by the combined action of RANBP1 and RANGAP1. In addition, the encoded protein may play a role both in apoptosis and in cell proliferation.Specifications
| CSE1L | |
| Polyclonal | |
| Unconjugated | |
| CSE1L | |
| CAS, CSE1, CSE1L, Exp2, Exportin 2, Importin alpha re exporter, XPO2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110750, 1434 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P55060, Q9ERK4 | |
| Cse1l | |
| CSE1L Fusion Protein Ag17556 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction