missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSE1L Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | CSE1L |
|---|---|
| Concentration | 0.19 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16851755
|
Proteintech
22219-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16841755
|
Proteintech
22219-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Proteins that carry a nuclear localization signal (NLS) are transported into the nucleus by the importin-alpha/beta heterodimer. Importin-alpha binds the NLS, while importin-beta mediates translocation through the nuclear pore complex. After translocation, RanGTP binds importin-beta and displaces importin-alpha. Importin-alpha must then be returned to the cytoplasm, leaving the NLS protein behind. The protein encoded by this gene binds strongly to NLS-free importin-alpha, and this binding is released in the cytoplasm by the combined action of RANBP1 and RANGAP1. In addition, the encoded protein may play a role both in apoptosis and in cell proliferation.Specifications
| CSE1L | |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P55060, Q9ERK4 | |
| 110750, 1434 | |
| CSE1L Fusion Protein Ag17556 | |
| Primary | |
| -20°C | |
| CSE1L |
| 0.19 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CAS, CSE1, CSE1L, Exp2, Exportin 2, Importin alpha re exporter, XPO2 | |
| Cse1l | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title