missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSPP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92596-0.1ml
This item is not returnable.
View return policy
Description
CSPP Polyclonal antibody specifically detects CSPP in Human, Mouse samples. It is validated for Western Blot
Specifications
| CSPP | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| Centrosome And Spindle Pole Associated Protein 1, Centrosome And Spindle Pole-Associated Protein 1, CSPP1, FLJ22490 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 962-1221 of human CSPP1 (NP_079066.5). QSPKGLDAATFQNVHDFNELKDRDSETRVDLKFMYLDPPRDHHTLEIQQQALLREQQKRLNRIKMQEGAKVDLDAIPSAKVREQRMPRDDTSDFLKNSLLESDSAFIGAYGETYPAIEDDVLPPPSQLPSARERRRNKWKGLDIDSSRPNVAPDGLSLKSISSVNVDELRVRNEERMRRLNEFHNKPINTDDESSLVDPDDIMKHIGDDGSNSVATEPWLRPGTSETLKRFMAEQLNQEQQQIPGKPGTFTWQGLSTAHG | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 79848 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction