missing translation for 'onlineSavingsMsg'
Learn More

CSPP Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 190.00 - € 463.00

Specifications

Antigen CSPP
Dilution Western Blot 1:500-1:1000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18649571
View Documents
Novus Biologicals
NBP2-92596-0.02ml
0.02 mL
€ 190.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18694362
View Documents
Novus Biologicals
NBP2-92596-0.1ml
0.1 mL
€ 463.00
0.01mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CSPP Polyclonal antibody specifically detects CSPP in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

CSPP
Western Blot
Unconjugated
Rabbit
Cell Biology, Cell Cycle and Replication
PBS with 50% glycerol, pH7.3.
79848
IgG
Affinity purified
Western Blot 1:500-1:1000
Polyclonal
Purified
RUO
Human, Mouse
Centrosome And Spindle Pole Associated Protein 1, Centrosome And Spindle Pole-Associated Protein 1, CSPP1, FLJ22490
Recombinant fusion protein containing a sequence corresponding to amino acids 962-1221 of human CSPP1 (NP_079066.5). QSPKGLDAATFQNVHDFNELKDRDSETRVDLKFMYLDPPRDHHTLEIQQQALLREQQKRLNRIKMQEGAKVDLDAIPSAKVREQRMPRDDTSDFLKNSLLESDSAFIGAYGETYPAIEDDVLPPPSQLPSARERRRNKWKGLDIDSSRPNVAPDGLSLKSISSVNVDELRVRNEERMRRLNEFHNKPINTDDESSLVDPDDIMKHIGDDGSNSVATEPWLRPGTSETLKRFMAEQLNQEQQQIPGKPGTFTWQGLSTAHG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.