missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTH Monoclonal Antibody (24GB5615)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA557375
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
CTH Monoclonal Antibody for Western Blot, Flow
This gene encodes a cytoplasmic enzyme in the trans-sulfuration pathway that converts cystathione derived from methionine into cysteine. Glutathione synthesis in the liver is dependent upon the availability of cysteine. Mutations in this gene cause cystathioninuria. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
Specifikationer
| CTH | |
| Monoclonal | |
| Unconjugated | |
| CTH | |
| 0610010I13Rik; AI314617; BC019483; CSE; Cth; cth protein; CTL target antigen; Cys3; cystathionase; cystathionase (cystathionine gamma-lyase); cystathionase isoform 1; cystathionine gamma-lyase; Cystathionine gamma-lyase-like protein; cysteine desulfhydrase; cysteine-protein sulfhydrase; EGK_00877; fb58d08; gamma-cystathionase; H; homoserine deaminase; homoserine dehydratase; LOC102117863; MGC9471; PRB-RA; probasin-related antigen; QccE-22305; synthase; Unknown (protein for MGC:85785); wu:fb48e03; wu:fb58d08; zgc:66001; zgc:85785 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 107869, 1491 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 24GB5615 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P32929, Q8VCN5 | |
| CTH | |
| Recombinant protein of human CTH (aa 1-405). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering