missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTH Monoclonal Antibody (24GB5615)

Mouse Monoclonal Antibody
€ 198.00 - € 564.00
Specifications
| Antigen | CTH |
|---|---|
| Clone | 24GB5615 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275375
![]() |
Invitrogen™
MA557375 |
100 μL |
€ 564.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275586
|
Invitrogen™
MA557376 |
20 μL |
€ 198.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CTH Monoclonal Antibody for Western Blot, Flow
This gene encodes a cytoplasmic enzyme in the trans-sulfuration pathway that converts cystathione derived from methionine into cysteine. Glutathione synthesis in the liver is dependent upon the availability of cysteine. Mutations in this gene cause cystathioninuria. Alternative splicing of this gene results in two transcript variants encoding different isoforms.Specifications
| CTH | |
| -20°C | |
| Monoclonal | |
| Liquid | |
| IgG2b | |
| Human, Mouse | |
| CTH | |
| 0610010I13Rik; AI314617; BC019483; CSE; Cth; cth protein; CTL target antigen; Cys3; cystathionase; cystathionase (cystathionine gamma-lyase); cystathionase isoform 1; cystathionine gamma-lyase; Cystathionine gamma-lyase-like protein; cysteine desulfhydrase; cysteine-protein sulfhydrase; EGK_00877; fb58d08; gamma-cystathionase; H; homoserine deaminase; homoserine dehydratase; LOC102117863; MGC9471; PRB-RA; probasin-related antigen; QccE-22305; synthase; Unknown (protein for MGC:85785); wu:fb48e03; wu:fb58d08; zgc:66001; zgc:85785 | |
| CTH | |
| Primary | |
| Affinity Chromatography |
| 24GB5615 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P32929, Q8VCN5 | |
| 107869, 1491 | |
| Recombinant protein of human CTH (aa 1-405). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title