Learn More
Invitrogen™ CTH Monoclonal Antibody (24GB5615)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA557376
 View more versions of this product
Product Code. 30275586
  • € 204.00 / 20µL
Description
CTH Monoclonal Antibody for Western Blot, Flow
This gene encodes a cytoplasmic enzyme in the trans-sulfuration pathway that converts cystathione derived from methionine into cysteine. Glutathione synthesis in the liver is dependent upon the availability of cysteine. Mutations in this gene cause cystathioninuria. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CTH
Monoclonal
Unconjugated
CTH
0610010I13Rik; AI314617; BC019483; CSE; Cth; cth protein; CTL target antigen; Cys3; cystathionase; cystathionase (cystathionine gamma-lyase); cystathionase isoform 1; cystathionine gamma-lyase; Cystathionine gamma-lyase-like protein; cysteine desulfhydrase; cysteine-protein sulfhydrase; EGK_00877; fb58d08; gamma-cystathionase; H; homoserine deaminase; homoserine dehydratase; LOC102117863; MGC9471; PRB-RA; probasin-related antigen; QccE-22305; synthase; Unknown (protein for MGC:85785); wu:fb48e03; wu:fb58d08; zgc:66001; zgc:85785
Mouse
Affinity Chromatography
RUO
107869, 1491
-20°C
Liquid
Flow Cytometry, Western Blot
24GB5615
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P32929, Q8VCN5
CTH
Recombinant protein of human CTH (aa 1-405).
20 μL
Primary
Human, Mouse
Antibody
IgG2b