missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Copper Nickel, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Copper Nickel, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Cadmium/Chromium/Aluminium/Manganese/Phosphorus/Silver/Arsenic/Cobalt/Tin/Lead/Iron/Nickel/Zinc/Copper |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cadmium 0.0021 %/Chromium 0.0029 %/Aluminium 0.007 %/Manganese 0.0095 %/Phosphorus 0.0299 %/Silver 0.042 %/Arsenic 0.047 %/Cobalt 0.0509 %/Tin 0.102 %/Lead 0.115 %/Iron 0.14 %/Nickel 14.38 %/Zinc 14.88 %/Copper 70.22 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction