missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Copper Nickel, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Copper Nickel, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Cadmium/Chromium/Silver/Arsenic/Cobalt/Zinc/Iron/Tin/Manganese/Lead/Phosphorus/Aluminium/Nickel/Copper |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cadmium 0.0021 %/Chromium 0.005 %/Silver 0.0064 %/Arsenic 0.0067 %/Cobalt 0.0079 %/Zinc 0.0203 %/Iron 0.0316 %/Tin 0.038 %/Manganese 0.0391 %/Lead 0.051 %/Phosphorus 0.053 %/Aluminium 1.95 %/Nickel 5.5 %/Copper 92.17 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction