Description
- Wide mouth, amber glass
- Pre-cleaned for visible particulates using deionised water
- With white polypropylene cap with PTFE faced foamed polyethylene liner
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Cleaning Procedure | USP purified water |
| Color | Amber |
| Material | Glass |
| Length (Metric) | 101mm |
| Quantity | 24/PK |
| Closure Color | White |
| Liner Material | PTFE |
| Product Type | Wide Mouth Bottle |
| Closure Liner | PTFE/Polyethylene |
| Show More |