missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B2 Antibody (X121.10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2612-0.025mg
This item is not returnable.
View return policy
Description
Cyclin B2 Monoclonal specifically detects Cyclin B2 in Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| Cyclin B2 | |
| Monoclonal | |
| 0.91 mg/ml | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| CCNB2 | |
| Xenopus laevis Cyclin B2 | |
| 0.025 mg | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 9133 | |
| Xenopus species | |
| Purified |
| Western Blot | |
| X121.10 | |
| Unconjugated | |
| cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Cyclin B2 (X121.10). Reacts with Xenopus Cyclin B2 only. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction