Learn More
Novus Biologicals™ Cyclin D3 Antibody (DCS2.2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-30977-100ug
 View more versions of this product
Product Code. 30066979
  • € 508.00 / 100µg
Description
Cyclin D3 Monoclonal antibody specifically detects Cyclin D3 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cyclin D3
Monoclonal
Unconjugated
cyclin D3, D3-type cyclin, G1/S-specific cyclin D3, G1/S-specific cyclin-D3
The immunogen for the mouse anti-human Cyclin D3 was a recombinant full length (34 kDa) Cyclin D3 protein.
100 μg
Cancer, Cell Cycle and Replication, Wnt Signaling Pathway
896
Store at -20°C. Avoid freeze/thaw cycles.
IgG1
Immunohistochemistry, Immunoprecipitation, Western Blot
DCS2.2
Phosphate buffered saline
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse, Rat