missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cyclin D3 Antibody (DCS2.2), Novus Biologicals™
Mouse Monoclonal Antibody
€ 213.00 - € 508.00
Specifications
| Antigen | Cyclin D3 |
|---|---|
| Clone | DCS2.2 |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067089
|
Novus Biologicals™
NBP3-30977-25ug |
25 μg |
€ 213.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066979
|
Novus Biologicals™
NBP3-30977-100ug |
100 μg |
€ 508.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin D3 Monoclonal antibody specifically detects Cyclin D3 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western BlotSpecifications
| Cyclin D3 | |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Wnt Signaling Pathway | |
| Phosphate buffered saline | |
| 896 | |
| IgG1 | |
| Protein A or G purified |
| DCS2.2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cyclin D3, D3-type cyclin, G1/S-specific cyclin D3, G1/S-specific cyclin-D3 | |
| The immunogen for the mouse anti-human Cyclin D3 was a recombinant full length (34 kDa) Cyclin D3 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title