missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclogalegenin, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Cyclogalegenin (Cyclogalegigenin) is a isoprenoid found in Astragalus galegiformis. Cyclogalegenin is the enantiomer of Cycloastragenol.
Specifications
Specifications
| Chemical Name or Material | Cyclogalegenin |
| CAS | 84605-18-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C30H50O5 |
| Quantity | 10 mg |
| Synonym | Cyclogalegigenin |
| Solubility Information | DMSO : 100 mg/mL (203.78 mM; Need ultrasonic) |
| SMILES | O[C@H]1CC[C@@]2(C3)[C@]([C@@H](O)C[C@]4([H])[C@@]23CC[C@@]5(C)[C@@]4(C)C[C@H](O)[C@]5([H])[C@@]6(C)O[C@@H](C(C)(C)O)CC6)([H])C1(C)C |
| Molecular Weight (g/mol) | 490.72 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur