missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 4A2, 4A3 Antibody (Clo4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50213
This item is not returnable.
View return policy
Description
Cytochrome P450 4A2, 4A3 Monoclonal antibody specifically detects Cytochrome P450 4A2, 4A3 in Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| Cytochrome P450 4A2, 4A3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, ELISA, Immunoprecipitation | |
| Clo4 | |
| Unconjugated | |
| PBS | |
| Rat liver cytochrome P450 4A2 and 4A3 | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction