missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 4A2, 4A3 Antibody (Clo4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00 - € 407.00
Specifications
| Antigen | Cytochrome P450 4A2, 4A3 |
|---|---|
| Clone | Clo4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654377
|
Novus Biologicals
NBP2-50213-0.025ml |
0.025 mL |
€ 190.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694367
|
Novus Biologicals
NBP2-50213 |
0.1 mL |
€ 407.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytochrome P450 4A2, 4A3 Monoclonal antibody specifically detects Cytochrome P450 4A2, 4A3 in Rat samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
| Cytochrome P450 4A2, 4A3 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| PBS | |
| IgG2a | |
| Protein A purified |
| Clo4 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Rat | |
| Rat liver cytochrome P450 4A2 and 4A3 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title