Learn More
Cytokeratin 1-specific Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-16848-100UL
 View more versions of this product
Product Code. 18067875
  • € 450.00 / 100µL
Description
The Cytokeratin 1-specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Cytokeratin 1-specific. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 1-specific antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
Cytokeratin 1-specific
1000 μg/mL
67 kDa cytokeratin, CK 1, CK1, Cytokeratin 1, EHK1, Hair alpha protein, K1, keratin 1, KRT1, KRT1 absorb, KRT1A, KRTA, Type II keratin Kb1
100 μL
3848
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa