Learn More
Cytokeratin 1-specific Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-16848-100UL
Description
The Cytokeratin 1-specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Cytokeratin 1-specific. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 1-specific antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| Cytokeratin 1-specific | |
| 1000 μg/mL | |
| 67 kDa cytokeratin, CK 1, CK1, Cytokeratin 1, EHK1, Hair alpha protein, K1, keratin 1, KRT1, KRT1 absorb, KRT1A, KRTA, Type II keratin Kb1 | |
| 100 μL | |
| 3848 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |