Learn More
Cytokeratin 1-specific Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | Cytokeratin 1-specific |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18067875
|
Proteintech
CL488-16848-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18045555
|
Proteintech
CL488-16848-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Cytokeratin 1-specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Cytokeratin 1-specific. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 1-specific antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| Cytokeratin 1-specific | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| 67 kDa cytokeratin, CK 1, CK1, Cytokeratin 1, EHK1, Hair alpha protein, K1, keratin 1, KRT1, KRT1 absorb, KRT1A, KRTA, Type II keratin Kb1 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 3848 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.