missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10 Antibody (DE-K10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47650-0.1MG
This item is not returnable.
View return policy
Description
Cytokeratin 10 Monoclonal specifically detects Cytokeratin 10 in Human, Canine, Feline samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cytokeratin 10 | |
| Monoclonal | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| KRT10 | |
| Cytoskeletal preparation extracted from human ectocervical epithelium | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This MAb recognizes a protein of 56.5kDa, identified as cytokeratin 10 (CK10). CK10 is expressed in all suprabasal layers of the epidermis. In the epidermis, expression of CK10 strictly parallels the extent of differentiation; it is absent in the basal layer, appears in the first suprabasal layers and increases in concentration towards the granular layer. However, CK10 is rarely detected in early stages of vulvar squamous carcinomas (tumors less than 2 cm, clinical stage I) regardless of the tumor grade. In larger and more advanced tumors (greater than 2 cm, clinical stages II and III), CK10 is detected very frequently. Expression of CK10 is related to maturation of malignant keratinocytes, being preferentially detected in more-differentiated parts. | |
| Store at 4C. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| DE-K10 | |
| Flow Cytometry 0.5-1ug/million cells in 0.1ml, Immunohistochemistry-Paraffin 0.1-0.2ug/ml, Immunofluorescence 0.5-1ug/ml | |
| BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
| Mouse | |
| 56.5 kDa | |
| 0.1 mg | |
| Cytoskeleton Markers | |
| 3858 | |
| Human, Canine, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction