Learn More
Cytokeratin 14 Rabbit, anti-Human, Clone: 1L19, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82824-1-RR-100UL
Description
The Cytokeratin 14 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Cytokeratin 14. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 14 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| Cytokeratin 14 | |
| 1L19 | |
| CK 14, CK14, Cytokeratin 14, EBS3, EBS4, K14, keratin 14, KRT14, NFJ | |
| 100 μL | |
| 3861 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 600 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |