Learn More
Cytokeratin 14 Rabbit, anti-Human, Clone: 1L19, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech 82824-1-RR-100UL
 View more versions of this product
Product Code. 18044305
  • € 439.00 / 100µL
Description
The Cytokeratin 14 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Cytokeratin 14. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 14 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
Cytokeratin 14
1L19
CK 14, CK14, Cytokeratin 14, EBS3, EBS4, K14, keratin 14, KRT14, NFJ
100 μL
3861
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, Immunohistochemistry, ELISA
600 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa