Learn More
Cytokeratin 14 Rabbit, anti-Human, Clone: 1L19, Proteintech
Rabbit Recombinant Monoclonal Antibody
€ 149.00 - € 439.00
Specifications
| Antigen | Cytokeratin 14 |
|---|---|
| Clone | 1L19 |
| Concentration | 600 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18044305
|
Proteintech
82824-1-RR-100UL |
100 μL |
€ 439.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18055205
|
Proteintech
82824-1-RR-20UL |
20 μL |
€ 149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Cytokeratin 14 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Cytokeratin 14. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 14 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| Cytokeratin 14 | |
| 600 μg/mL | |
| Liquid | |
| Human, Mouse, Rat | |
| CK 14, CK14, Cytokeratin 14, EBS3, EBS4, K14, keratin 14, KRT14, NFJ | |
| Primary |
| 1L19 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Rabbit | |
| 3861 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.