Learn More
Cytokeratin 7, 8 Antibody (CAM5.2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44630-100ug
 View more versions of this product
Product Code. 30288041
  • € 484.00 / 100µg
Description
Cytokeratin 7, 8 Monoclonal antibody specifically detects Cytokeratin 7, 8 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytokeratin 7, 8
Monoclonal
0.2 mg/ml
10 mM PBS with 0.05% BSA
Mouse
Protein A purified
RUO
Primary
Human
Purified
Immunohistochemistry
CAM5.2
Unconjugated
CK7, CK-7, CK8, CK-8, Cytokeratin-7, Cytokeratin-8, keratin 8, keratin, type II cytoskeletal 7, keratin-7
Human colorectal carcinoma cell line HT29
100 μg
Cellular Signaling
3855
Store at 4⪚C.
IgG2a κ