missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus gH Antibody (HCMV16) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20158-0.2mg
This item is not returnable.
View return policy
Description
Cytomegalovirus gH Monoclonal antibody specifically detects Cytomegalovirus gH in Cytomegalovirus, Virus samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Neutralization
Specifications
| Cytomegalovirus gH | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3077450 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG κ |
| ELISA, Immunofluorescence, Immunoprecipitation, Neutralization | |
| HCMV16 | |
| Unconjugated | |
| envelope glycoprotein H, gH, gpUL75, HCMV gH, HHV-5 gH, HHV5 glycoprotein H, Human betaherpesvirus 5 envelope glycoprotein H, Human Cytomegalovirus glycoprotein H, UL75 | |
| The original antibody was generated by immunization of mice with purified HCMV strain AD169. | |
| 0.2 mg | |
| Primary | |
| Cytomegalovirus (CMV), Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction