missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus gH Antibody (HCMV16) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 267.00 - € 635.00
Specifications
| Antigen | Cytomegalovirus gH |
|---|---|
| Clone | HCMV16 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunofluorescence, Immunoprecipitation, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610355
|
Novus Biologicals
NBP3-20158-0.025mg |
0.025 mg |
€ 267.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674714
|
Novus Biologicals
NBP3-20158-0.2mg |
0.2 mg |
€ 635.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytomegalovirus gH Monoclonal antibody specifically detects Cytomegalovirus gH in Cytomegalovirus, Virus samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, NeutralizationSpecifications
| Cytomegalovirus gH | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 3077450 | |
| IgG κ | |
| Protein A purified |
| HCMV16 | |
| ELISA, Immunofluorescence, Immunoprecipitation, Neutralization | |
| Unconjugated | |
| Rabbit | |
| Cytomegalovirus (CMV), Virus | |
| envelope glycoprotein H, gH, gpUL75, HCMV gH, HHV-5 gH, HHV5 glycoprotein H, Human betaherpesvirus 5 envelope glycoprotein H, Human Cytomegalovirus glycoprotein H, UL75 | |
| The original antibody was generated by immunization of mice with purified HCMV strain AD169. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title