missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Dimer Antibody (3B6) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08986-0.025mg
This item is not returnable.
View return policy
Description
D-Dimer Monoclonal antibody specifically detects D-Dimer in Human samples. It is validated for ELISA, Lateral Flow, Agglutination
Specifications
| D-Dimer | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
| ELISA, Lateral Flow, Agglutination | |
| 3B6 | |
| Unconjugated | |
| D Dimer | |
| Human cross-linked D-Dimer. | |
| 0.025 mg | |
| Extracellular Matrix | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction