missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Dimer Antibody (3B6) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 250.00 - € 705.00
Specifications
| Antigen | D-Dimer |
|---|---|
| Clone | 3B6 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Lateral Flow, Agglutination |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627606
|
Novus Biologicals
NBP3-08986-0.2mg |
0.2 mg |
€ 705.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625466
|
Novus Biologicals
NBP3-08986-0.025mg |
0.025 mg |
€ 250.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D-Dimer Monoclonal antibody specifically detects D-Dimer in Human samples. It is validated for ELISA, Lateral Flow, AgglutinationSpecifications
| D-Dimer | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| D Dimer | |
| IgG κ | |
| Protein A purified |
| 3B6 | |
| ELISA, Lateral Flow, Agglutination | |
| Unconjugated | |
| Rabbit | |
| Extracellular Matrix | |
| PBS | |
| Human cross-linked D-Dimer. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title