missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abhd5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 26 publications
Brand: Novus Biologicals NB110-41576
This item is not returnable.
View return policy
Description
Abhd5 Polyclonal specifically detects Abhd5 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.
Specifications
| Abhd5 | |
| Polyclonal | |
| Western Blot 1:500, Simple Western 1:1000 | |
| Q8WTS1 | |
| ABHD5 | |
| A synthetic peptide made to an internal region within residues 200-300 of the human Abhd5 protein. [Swiss-Prot# Q8WTS1] | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
| 51099 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| abhydrolase domain containing 5, Abhydrolase domain-containing protein 5,1-acylglycerol-3-phosphate O-acyltransferase ABHD5, CDS, CGI58, EC 2.3.1.51, IECN2, Lipid droplet-binding protein CGI-58, MGC8731, NCIE2CGI-58 | |
| Rabbit | |
| IgG purified | |
| RUO | |
| Primary | |
| Human and rat. Mouse reactivity reported in scientific literature (PMID:23878361). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction