missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP Kinase 2 Antibody (S09-8F2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19618-100ul
This item is not returnable.
View return policy
Description
DAP Kinase 2 Monoclonal antibody specifically detects DAP Kinase 2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| DAP Kinase 2 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S09-8F2 | |
| Unconjugated | |
| DAP kinase 2, DAP-kinase-related protein 1, DAP-kinase-related protein 1 beta isoform, death-associated protein kinase 2, DRP1, DRP-1MGC119312, EC 2.7.11, EC 2.7.11.1 | |
| A synthetic peptide of human DAP Kinase 2 (Uniprot # Q9UIK4) | |
| 100 μg | |
| Apoptosis, Autophagy, Cellular Markers | |
| 23604 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction