Learn More
Medline Scientific™ Darkfield Condenser
Used to observe low contrast samples such as live blood samples and available in both dry and Cardioid (oil) variants.
Brand:  Medline Scientific™ 3132.5201
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11980279
  • € 347.00 / Each
Estimated Shipment: 24-07-2024
to see stock.
Add to basket
Description
Description
  • For use with Triton II Fluorescence Microscopes
  • Dry darkfield for NA 0.70 – 0.90
  • Cardioid darkfield for NA 1.25 - 1.36
Specifications
Specifications
Darkfield Dry
Low contrast sample viewing
Triton II Fluorescence microscope