Learn More
Medline Scientific™ Darkfield Condenser
Used to observe low contrast samples such as live blood samples and available in both dry and Cardioid (oil) variants.
€ 347.00 - € 456.00
Specifications
For Use With (Application) Low contrast sample viewing
For Use With (Equipment) Triton II Fluorescence microscope
View More Specs
Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
11980279
View Documents Promotion Details
Medline Scientific™
3132.5201
Darkfield Dry
€ 347.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
11990279
View Documents Promotion Details
Medline Scientific™
3133.5322
Darkfield Cardioid Oil
€ 456.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
  • For use with Triton II Fluorescence Microscopes
  • Dry darkfield for NA 0.70 – 0.90
  • Cardioid darkfield for NA 1.25 - 1.36
Specifications
Specifications
Low contrast sample viewing
Triton II Fluorescence microscope