Learn More
Medline Scientific™ Darkfield Condenser
Used to observe low contrast samples such as live blood samples and available in both dry and Cardioid (oil) variants.
€ 347.00 - € 456.00
Specifications
For Use With (Application) | Low contrast sample viewing |
---|---|
For Use With (Equipment) | Triton II Fluorescence microscope |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11980279
|
Medline Scientific™
3132.5201 |
Darkfield Dry |
€ 347.00
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
11990279
|
Medline Scientific™
3133.5322 |
Darkfield Cardioid Oil |
€ 456.00
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
Description
- For use with Triton II Fluorescence Microscopes
- Dry darkfield for NA 0.70 – 0.90
- Cardioid darkfield for NA 1.25 - 1.36
Specifications
Low contrast sample viewing | |
Triton II Fluorescence microscope |