Learn More
Medline Scientific™ Darkfield Condenser
Used to observe low contrast samples such as live blood samples and available in both dry and Cardioid (oil) variants.
Brand: Medline Scientific™ 3133.5322
Additional Details : Weight : 0.01000kg
Description
- For use with Triton II Fluorescence Microscopes
- Dry darkfield for NA 0.70 – 0.90
- Cardioid darkfield for NA 1.25 - 1.36
Specifications
Darkfield Cardioid Oil | |
Low contrast sample viewing | |
Triton II Fluorescence microscope |