missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCPS Monoclonal Antibody (OTI4H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505626
This item is not returnable.
View return policy
Description
Necessary for the complete degradation of mRNAs, both in normal mRNA turnover and in nonsense-mediated mRNA decay. Removes the 7-methyl guanine cap structure from mRNA fragments shorter than 10 nucleotides that are produced by 3′->5′ exosome-mediated mRNA decay. Releases m7GMP. Can also degrade m7GDP to m7GMP. Has no activity towards mRNA molecules longer than 25 nucleotides.Specifications
| DCPS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96C86 | |
| DCPS | |
| Full length human recombit protein of human DCPS produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4H8 | |
| Unconjugated | |
| DCPS | |
| ARS, DCS1, HINT-5, HINT5, HSL1, HSPC015 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 28960 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction