missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dectin 1 (soluble) Monoclonal Antibody (2A11), PE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA516483

 View more versions of this product

Product Code. 15225789

  • € 266.00 / 25 tests

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. Once reconstituted, this product has a shelf life of approx. 18 months. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse Dectin-1 antibody, clone 2A11 recognizes murine beta-glucan receptor, also known as Dectin-1. Rat anti Mouse Dectin-1 antibody, clone 2A11 recognizes murine beta-glucan receptor, also known as Dectin-1.

CLEC7A, also known as dectin-1, is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily and is predominantly expressed on myeloid cells. It is a small glycoprotein type II membrane receptor with an extracellular C-type lectin-like domain fold and a cytoplasmic domain with an immunoreceptor tyrosine-based activation motif (ITAM). CLEC7A functions as a pattern-recognition receptor that recognizes a variety of beta-1,3-linked and beta-1,6-linked glucans from fungi and plants, and in this way plays a role in innate immune response. Upon fungal exposure, CLEC7A activates Syk tyrosine kinase, triggering a massive oxidative burst through the formation of reactive oxygen species.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dectin 1 (soluble)
Monoclonal
Conc. Not Determined
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
Q6QLQ4
Clec7a
Dectin-1 transfected NIH3T3 cells and recombinant soluble Dectin-1.
25 Tests
Primary
Mouse
Antibody
IgG2b
Flow Cytometry
2A11
PE
Clec7a
Beta-glucan receptor; beta-GR; BGR; CANDF4; CD369; CLEC7A; Clec7b; CLECSF12; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 12; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 12; C-type lectin domain family 7 member A; C-type lectin domain family 7, member A; C-type lectin superfamily member 12; DC-associated C-type lectin 1; DECTIN1; dectin-1; Dendritic cell-associated C-type lectin 1; dendritic cell-associated C-type lectin-1; lectin-like receptor 1; RGD1565140; SCARE2; UNQ539/PRO1082
Rat
Protein G
RUO
56644
4°C, store in dark, DO NOT FREEZE!
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.