Learn More
Invitrogen™ Dectin 1 (soluble) Monoclonal Antibody (2A11), PE

Rat Monoclonal Antibody
€ 255.00 - € 552.00
Specifications
| Antigen | Dectin 1 (soluble) |
|---|---|
| Clone | 2A11 |
| Concentration | Conc. Not Determined |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15215789
![]() |
Invitrogen™
MA516482 |
100 Tests |
€ 552.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15225789
|
Invitrogen™
MA516483 |
25 Tests |
€ 255.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. Once reconstituted, this product has a shelf life of approx. 18 months. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse Dectin-1 antibody, clone 2A11 recognizes murine beta-glucan receptor, also known as Dectin-1. Rat anti Mouse Dectin-1 antibody, clone 2A11 recognizes murine beta-glucan receptor, also known as Dectin-1.
CLEC7A, also known as dectin-1, is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily and is predominantly expressed on myeloid cells. It is a small glycoprotein type II membrane receptor with an extracellular C-type lectin-like domain fold and a cytoplasmic domain with an immunoreceptor tyrosine-based activation motif (ITAM). CLEC7A functions as a pattern-recognition receptor that recognizes a variety of beta-1,3-linked and beta-1,6-linked glucans from fungi and plants, and in this way plays a role in innate immune response. Upon fungal exposure, CLEC7A activates Syk tyrosine kinase, triggering a massive oxidative burst through the formation of reactive oxygen species.Specifications
| Dectin 1 (soluble) | |
| Conc. Not Determined | |
| Flow Cytometry | |
| PE | |
| Rat | |
| RUO | |
| PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
| Q6QLQ4 | |
| 56644 | |
| Dectin-1 transfected NIH3T3 cells and recombinant soluble Dectin-1. | |
| Antibody |
| 2A11 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Lyophilized | |
| IgG2b | |
| Mouse | |
| Clec7a | |
| Beta-glucan receptor; beta-GR; BGR; CANDF4; CD369; CLEC7A; Clec7b; CLECSF12; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 12; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 12; C-type lectin domain family 7 member A; C-type lectin domain family 7, member A; C-type lectin superfamily member 12; DC-associated C-type lectin 1; DECTIN1; dectin-1; Dendritic cell-associated C-type lectin 1; dendritic cell-associated C-type lectin-1; lectin-like receptor 1; RGD1565140; SCARE2; UNQ539/PRO1082 | |
| Clec7a | |
| Primary | |
| Protein G |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.