missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmocollin-2 Rabbit anti-Rat, Clone: 3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90904-100UL
This item is not returnable.
View return policy
Description
Desmocollin-2 Monoclonal specifically detects Desmocollin-2 in Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Desmocollin-2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1824 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 3 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:250-1:1000 | |
| ARVD11, Cadherin family member 2, CDHF2DGII/III, desmocollin 2, desmocollin-2, Desmocollin-3, Desmosomal glycoprotein II, desmosomal glycoprotein II/III, Desmosomal glycoprotein III, DG2, DSC3DKFZp686I11137 | |
| Recombinant Rat Desmocollin-2 Protein | |
| 100 μg | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction