missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmocollin-2 Rabbit anti-Rat, Clone: 3, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 367.00 - € 552.00
Specifications
| Antigen | Desmocollin-2 |
|---|---|
| Clone | 3 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:250-1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18009356
|
Novus Biologicals
NBP2-90904-50UL |
50 μg |
€ 367.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18056487
|
Novus Biologicals
NBP2-90904-100UL |
100 μg |
€ 552.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Desmocollin-2 Monoclonal specifically detects Desmocollin-2 in Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Desmocollin-2 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:250-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 1824 | |
| IgG | |
| Protein A purified |
| 3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| ARVD11, Cadherin family member 2, CDHF2DGII/III, desmocollin 2, desmocollin-2, Desmocollin-3, Desmosomal glycoprotein II, desmosomal glycoprotein II/III, Desmosomal glycoprotein III, DG2, DSC3DKFZp686I11137 | |
| Recombinant Rat Desmocollin-2 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title