missing translation for 'onlineSavingsMsg'
Learn More

Desoxybenzoin p-toluenesulfonylhydrazone, 97%, Thermo Scientific™

Product Code. 15450268
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450268 5 g 5g
15460268 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450268 Supplier Thermo Scientific Alfa Aesar Supplier No. H53447.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Desoxybenzoin p-toluenesulfonylhydrazone is used as the reagent.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19816-85-4
Molecular Formula C21H20N2O2S
Molecular Weight (g/mol) 364.463
MDL Number MFCD06411289
InChI Key INMYHVHTZKDKDK-DQRAZIAOSA-N
Synonym desoxybenzoin p-toluenesulfonylhydrazone, n-e-1,2-diphenylethylideneamino-4-methylbenzenesulfonamide, toluene-4-sulfonic acid dibenzyl-alpha-ylidenehydrazone, n'-1z-1,2-diphenylethylidene-4-methylbenzenesulfonohydrazide
PubChem CID 5834443
IUPAC Name N-[(Z)-1,2-diphenylethylideneamino]-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NN=C(CC2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 139°C to 143°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 364.46
Percent Purity 97%
Chemical Name or Material Desoxybenzoin p-toluenesulfonylhydrazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.