missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DGKZ Monoclonal Antibody (K49027_8F3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555685
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: STGSKDVVRY LLDHAPPEIL DAVEENGETC LHQAAALGQR TICHYIVEAG ASLMKTDQQG DTPRQRAEKA QDTELAAYLE NRQHYQMIQR
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It may attenuate protein kinase C activity by regulating diacylglycerol levels in intracellular signaling cascade and signal transduction. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified.
Specifications
| DGKZ | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q13574, Q80UP3 | |
| DGKZ | |
| Recombinant human DGKZ (immunogen range is 832-921). | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| K49027_8F3 | |
| Unconjugated | |
| DGKZ | |
| 104 kDa diacylglycerol kinase; DAG kinase DAGK6; DAG kinase zeta; DAGK5; DAGK6; DGK-IV; DGKZ; DGK-zeta; Diacylglycerol kinase zeta; diacylglycerol kinase, zeta 104kDa; Diglyceride kinase; diglyceride kinase zeta; E130307B02Rik; ENSMUSG00000075035; F730209L11Rik; hDGKzeta; mDGK[z] | |
| Mouse | |
| Protein A | |
| RUO | |
| 104418, 8525 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction