missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DGKZ Monoclonal Antibody (K49027_8F3), Invitrogen™
Mouse Monoclonal Antibody
€ 493.00 - € 868.00
Specifications
| Antigen | DGKZ |
|---|---|
| Clone | K49027_8F3 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30199347
|
Invitrogen™
MA555685 |
50 μL |
€ 493.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30211914
|
Invitrogen™
MA556065 |
200 μL |
€ 868.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: STGSKDVVRY LLDHAPPEIL DAVEENGETC LHQAAALGQR TICHYIVEAG ASLMKTDQQG DTPRQRAEKA QDTELAAYLE NRQHYQMIQR
DGKZ Monoclonal antibody specifically detects DGKZ in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| DGKZ | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 104 kDa diacylglycerol kinase; DAG kinase DAGK6; DAG kinase zeta; DAGK5; DAGK6; DGK-IV; DGKZ; DGK-zeta; Diacylglycerol kinase zeta; diacylglycerol kinase, zeta 104kDa; Diglyceride kinase; diglyceride kinase zeta; E130307B02Rik; ENSMUSG00000075035; F730209L11Rik; hDGKzeta; mDGK[z] | |
| Recombinant human DGKZ (immunogen range is 832-921). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| DGKZ |
| K49027_8F3 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q13574, Q80UP3 | |
| 104418, 8525 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title