missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyldichlorosilane, 97%

Product Code. 11369218
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11369218

Brand: Thermo Scientific Alfa Aesar 042599.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18416-07-4 | C16H34Cl2Si | 325.433 g/mol

Di-n-octyldichlorosilane is used in the synthesis of organic photovoltaics. It is also used in medicine field, electronic industry and polymer material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18416-07-4
Molecular Formula C16H34Cl2Si
Molecular Weight (g/mol) 325.433
MDL Number MFCD01074472
InChI Key CVAGYCLEIYGJQT-UHFFFAOYSA-N
Synonym di-n-octyldichlorosilane, dichloro dioctyl silane, silane, dichlorodioctyl, dioctyldichlorosilane, dioctyldichlorsilan, dichlorodi-n-octylsilane, di-n-octyldichlorosilane 10g
PubChem CID 248429
IUPAC Name dichloro(dioctyl)silane
SMILES CCCCCCCC[Si](CCCCCCCC)(Cl)Cl

Specifications

Boiling Point 145°C (0.2mmHg)
Color Colorless to Yellow
Physical Form Liquid
Quantity 50 g
Linear Formula [CH3(CH2)7]2SiCl2
UN Number UN2987
Solubility Information Sparingly Soluble (8.8E-6g/L) at 25°C.
Formula Weight 325.44
Percent Purity 97%
Sensitivity Moisture sensitive
Density 0.940 g/mL
Chemical Name or Material Di-n-octyldichlorosilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.