missing translation for 'onlineSavingsMsg'
Learn More

Dibromo(1,5-cyclooctadiene)platinum(II), Pt 41.6% min, Thermo Scientific™

Product Code. 11343688
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343688 5 g 5g
11353688 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343688 Supplier Thermo Scientific Alfa Aesar Supplier No. 039282.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12145-48-1
Molecular Formula C8H12Br2Pt
Molecular Weight (g/mol) 463.08
MDL Number MFCD00058724
InChI Key HFLCCHAOKSABAA-PHFPKPIQSA-L
Synonym dibromo 1,5-cyclooctadiene platinum ii, c8h12.br2pt, 1z,5z-cycloocta-1,5-diene; dibromoplatinum, 1,5-cyclooctadiene platinum ii bromide, dibromo cycloocta-1,5-diene platinum ii, 1,5-cyclooctadiene, z,z-; dibromoplatinum
PubChem CID 92135605
SMILES [Br-].[Br-].[Pt++].C1C\C=C/CC\C=C/1

Specifications

Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;platinum(2+);dibromide
Formula Weight 463.09
Concentration or Composition (by Analyte or Components) Pt 41.6% min.
Physical Form Powder
Chemical Name or Material Dibromo(1,5-cyclooctadiene)platinum(II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.