missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibromo-tyrosine Antibody (9F12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59351-25ug
This item is not returnable.
View return policy
Description
Dibromo-tyrosine Monoclonal specifically detects Dibromo-tyrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Dibromo-tyrosine | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide | |
| Synthetic 3,5-Dibromotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| 25 μg | |
| Primary | |
| Store at -20°C. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 9F12 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Specific for 3,5-Dibromo-tyrosine. Does not cross-react with Nitrotyrosine. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction