missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibromo-tyrosine Antibody (9F12), Novus Biologicals™
Mouse Monoclonal Antibody
€ 178.00 - € 600.00
Specifications
| Antigen | Dibromo-tyrosine |
|---|---|
| Clone | 9F12 |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
Description
Dibromo-tyrosine Monoclonal specifically detects Dibromo-tyrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Dibromo-tyrosine | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 | |
| Protein G purified |
| 9F12 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Synthetic 3,5-Dibromotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| Primary | |
| Specific for 3,5-Dibromo-tyrosine. Does not cross-react with Nitrotyrosine. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title