missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethane, 99.7%, Honeywell™

Product Code. p-7120377 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
2 L
2.5 L
Packaging:
Glass bottle
Unit Size:
2 L
500mL
This item is not returnable. View return policy

Product Code. 15655810

Brand: Honeywell Research Chemicals 5382802L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Contains 50-150 ppm BHT as stabilizer, 99.7%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-95-3
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00009246
InChI Key KLKFAASOGCDTDT-UHFFFAOYSA-N
Synonym diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene
PubChem CID 10024
IUPAC Name (ethoxymethoxy)ethane
SMILES CCOCOCC

Specifications

Melting Point -50°C
Color Colorless
Density 0.831g/mL at 25°C
Boiling Point 255°C
Packaging Glass bottle
Linear Formula CH2(OC2H5)2
Quantity 2 L
UN Number UN2373
Beilstein 1697253
Vapor Density 3.6 (vs air)
Formula Weight 104.15g/mol
Vapor Pressure 60mmHg (25°C)
Percent Purity 0.97
Chemical Name or Material Diethoxymethane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.