missing translation for 'onlineSavingsMsg'
Learn More

Difloxacin hydrochloride, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J63754.14

Quantity 25g

 View more versions of this product

Product Code. 15405939

  • ONLINE EXCLUSIVE PRICE
    € 422.00
    € 414.02 / 25g
    Save € 7.98
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • A quinolone antibacterial compound
  • Structurally related to norfloxacin
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

91296-86-5
435.856
JFMGBGLSDVIOHL-UHFFFAOYSA-N
56205
CN1CCN(CC1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl
C21H20ClF2N3O3
MFCD03840489
difloxacin hydrochloride, difloxacin hcl, unii-xj0260hj0o, abbott-56619, 6-fluoro-1-4-fluorophenyl-7-4-methylpiperazin-1-yl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, difloxacin hydrochloride usan, dsstox_cid_26599, dsstox_rid_81754, dsstox_gsid_46599
6-fluoro-1-(4-fluorophenyl)-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride

Specifications

91296-86-5
MFCD03840489
6464798
difloxacin hydrochloride, difloxacin hcl, unii-xj0260hj0o, abbott-56619, 6-fluoro-1-4-fluorophenyl-7-4-methylpiperazin-1-yl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, difloxacin hydrochloride usan, dsstox_cid_26599, dsstox_rid_81754, dsstox_gsid_46599
JFMGBGLSDVIOHL-UHFFFAOYSA-N
6-fluoro-1-(4-fluorophenyl)-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride
56205
Difloxacin hydrochloride
C21H20ClF2N3O3
25g
14,3141
Soluble in aqueous buffer
CN1CCN(CC1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl
435.856
435.85
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.