Learn More
Difloxacin hydrochloride, Thermo Scientific™
€ 126.00 - € 414.02
Chemical Identifiers
| CAS | 91296-86-5 |
|---|---|
| Molecular Formula | C21H20ClF2N3O3 |
| Molecular Weight (g/mol) | 435.856 |
| MDL Number | MFCD03840489 |
| InChI Key | JFMGBGLSDVIOHL-UHFFFAOYSA-N |
| Synonym | difloxacin hydrochloride, difloxacin hcl, unii-xj0260hj0o, abbott-56619, 6-fluoro-1-4-fluorophenyl-7-4-methylpiperazin-1-yl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, difloxacin hydrochloride usan, dsstox_cid_26599, dsstox_rid_81754, dsstox_gsid_46599 |
| PubChem CID | 56205 |
| IUPAC Name | 6-fluoro-1-(4-fluorophenyl)-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride |
| SMILES | CN1CCN(CC1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15495929
|
Thermo Scientific Alfa Aesar
J63754.06 |
5g |
€ 129.00 € 126.00 / 5g Save € 3.00 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15405939
|
Thermo Scientific Alfa Aesar
J63754.14 |
25g |
€ 422.00 € 414.02 / 25g Save € 7.98 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- A quinolone antibacterial compound
- Structurally related to norfloxacin
Chemical Identifiers
| 91296-86-5 | |
| 435.856 | |
| JFMGBGLSDVIOHL-UHFFFAOYSA-N | |
| 56205 | |
| CN1CCN(CC1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl |
| C21H20ClF2N3O3 | |
| MFCD03840489 | |
| difloxacin hydrochloride, difloxacin hcl, unii-xj0260hj0o, abbott-56619, 6-fluoro-1-4-fluorophenyl-7-4-methylpiperazin-1-yl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, difloxacin hydrochloride usan, dsstox_cid_26599, dsstox_rid_81754, dsstox_gsid_46599 | |
| 6-fluoro-1-(4-fluorophenyl)-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride |
Specifications
| 91296-86-5 | |
| MFCD03840489 | |
| 14,3141 | |
| Soluble in aqueous buffer | |
| CN1CCN(CC1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl | |
| 435.856 | |
| 435.85 |
| C21H20ClF2N3O3 | |
| 6464798 | |
| difloxacin hydrochloride, difloxacin hcl, unii-xj0260hj0o, abbott-56619, 6-fluoro-1-4-fluorophenyl-7-4-methylpiperazin-1-yl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, difloxacin hydrochloride usan, dsstox_cid_26599, dsstox_rid_81754, dsstox_gsid_46599 | |
| JFMGBGLSDVIOHL-UHFFFAOYSA-N | |
| 6-fluoro-1-(4-fluorophenyl)-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride | |
| 56205 | |
| Difloxacin hydrochloride |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.